Learn More
Globe Scientific Oak Ridge Style Centrifuge Tubes
Catalog No. 21402931
Change view
Click to view available options
Capacity (Metric):
10 mL
30 mL
50 mL
70 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-402-931 10 mL
21-402-932 30 mL
21-402-933 50 mL
21-402-934 70 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-931 Supplier Globe Scientific Supplier No. 6320
Add to Cart
Add to Cart
Economic choice for high speed refrigerated or non-refrigerated centrifugation, designed with leakproof linerless closures
  • Molded from durable, chemical resistant polypropylene, the tubes have round bottoms and consistent wall thicknesses for centrifugation at up to 50000 g when filled to 80% of their nominal capacity
  • Oakridge style tubes are autoclavable and re-usable and are designed with leakproof linerless closures
  • Oakridge Style Centrifuge Tubes Features:
    • Tubes have round bottoms and consistent wall thickness for high speed refrigerated or non-refrigerated centrifugation
    • Tubes withstand up to 50000 g when filled to 80% of volume - Tubes must be filled to 80% of volume for proper performance
    • Re-usable and autoclavable at 121°C
    • Round bottoms fit high speed centrifuge rotors
    • Polypropylene (PP) tube and screw cap with integral seal ring for leakproof closure
    • Excellent chemical resistance
Specifications
Capacity (Metric) 10 mL
Disposable Reusable
Material Polypropylene
Sterility Non-sterile
Autoclavable Autoclavable
Shape Cylindrical
Bottom Shape Round
Closure Type Attached Screw Cap
Closure Included Yes
Writing Area No
Graduated No
Product Type Centrifuge Tube
Max. RCF 50,000 x g