Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top
SCI_ED

Catalog No. S48328 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 48 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 48 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
Height (English):
34 in.
36 in.
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English)
S48328 36 x 54 in. 36 in.
S41918 24 x 72 in. 34 in.
S42117 24 x 60 in. 34 in.
S42709 30 x 48 in. 34 in.
S47464 24 x 48 in. 34 in.
S42749 30 x 60 in. (76.2 x 152.4 cm) 34 in.
S47620 30 x 48 in. 36 in.
S42124 24 x 60 in. 36 in.
S47690 30 x 54 in. 34 in.
S48072 21 x 28 in. 34 in.
S47697 30 x 54 in. 36 in.
S47824 30 x 60 in. (76.2 x 152.4 cm) 36 in.
S48157 36 x 48 in. 34 in.
S47970 30 x 72 in. 34 in.
S48439 24 x 54 in. 34 in.
S48243 21 x 54 in. 36 in.
S48321 36 x 54 in. 34 in.
S48446 24 x 54 in. 36 in.
S48566 21 x 60 in. 34 in.
S48573 21 x 60 in. 36 in.
S41822 36 x 72 in. 34 in.
S48719 21 x 72 in. 34 in.
S48651 36 x 60 in. 34 in.
S48658 36 x 60 in. 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. S48328 Supplier Diversified Spaces™ Supplier No. P7194K36N
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Dimensions (L x W) 36 x 54 in.
Certifications/Compliance SEFA compliant and MAS Certified Green™
Load Bearing Capacity (Metric) 226.79 kg
Top Style Solid Phenolic
Style Plain Apron
Adjustable No
Load Bearing Capacity (English) 500 lb.
For Use With (Application) High school and above and with low temperature use
Finish Chemical resistant, earth-friendly UV
Height (English) 36 in.
Height (Metric) 91.44 cm
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.