Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Leg Brace and Electrical Option
SCI_ED

Catalog No. S42744 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 48 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 48 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
42 x 48 in.
42 x 54 in.
42 x 60 in.
42 x 72 in.
Height (English):
30 in.
34 in.
36 in.
Includes:
Leg Brace, UL-rated Electrical
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English) Includes
S42744 30 x 60 in. (76.2 x 152.4 cm) 30 in. Leg Brace, UL-rated Electrical
S41910 24 x 72 in. 30 in. Leg Brace, UL-rated Electrical
S41998 42 x 72 in. 30 in. Leg Brace, UL-rated Electrical
S42116 24 x 60 in. 34 in. Leg Brace, UL-rated Electrical
S42002 42 x 72 in. 34 in. Leg Brace, UL-rated Electrical
S41917 24 x 72 in. 34 in. Leg Brace, UL-rated Electrical
S42123 24 x 60 in. 36 in. Leg Brace, UL-rated Electrical
S42228 42 x 54 in. 34 in. Leg Brace, UL-rated Electrical
S42223 42 x 54 in. 30 in. Leg Brace, UL-rated Electrical
S42286 42 x 60 in. 34 in. Leg Brace, UL-rated Electrical
S42291 42 x 60 in. 36 in. Leg Brace, UL-rated Electrical
S42708 30 x 48 in. 34 in. Leg Brace, UL-rated Electrical
S47458 24 x 48 in. 30 in. Leg Brace, UL-rated Electrical
S42787 36 x 54 in. 34 in. Leg Brace, UL-rated Electrical
S47463 24 x 48 in. 34 in. Leg Brace, UL-rated Electrical
S47468 24 x 48 in. 36 in. Leg Brace, UL-rated Electrical
S47619 30 x 48 in. 36 in. Leg Brace, UL-rated Electrical
S47609 30 x 48 in. 30 in. Leg Brace, UL-rated Electrical
S47682 30 x 54 in. 30 in. Leg Brace, UL-rated Electrical
S47552 42 x 48 in. 34 in. Leg Brace, UL-rated Electrical
S47689 30 x 54 in. 34 in. Leg Brace, UL-rated Electrical
S47696 30 x 54 in. 36 in. Leg Brace, UL-rated Electrical
S47823 30 x 60 in. (76.2 x 152.4 cm) 36 in. Leg Brace, UL-rated Electrical
S47962 30 x 72 in. 30 in. Leg Brace, UL-rated Electrical
S48064 21 x 28 in. 30 in. Leg Brace, UL-rated Electrical
S47969 30 x 72 in. 34 in. Leg Brace, UL-rated Electrical
S48071 21 x 28 in. 34 in. Leg Brace, UL-rated Electrical
S48156 36 x 48 in. 34 in. Leg Brace, UL-rated Electrical
S48149 36 x 48 in. 30 in. Leg Brace, UL-rated Electrical
S48235 21 x 54 in. 34 in. Leg Brace, UL-rated Electrical
S48163 36 x 48 in. 36 in. Leg Brace, UL-rated Electrical
S48228 21 x 54 in. 30 in. Leg Brace, UL-rated Electrical
S48327 36 x 54 in. 36 in. Leg Brace, UL-rated Electrical
S48313 36 x 54 in. 30 in. Leg Brace, UL-rated Electrical
S48438 24 x 54 in. 34 in. Leg Brace, UL-rated Electrical
S48445 24 x 54 in. 36 in. Leg Brace, UL-rated Electrical
S48565 21 x 60 in. 34 in. Leg Brace, UL-rated Electrical
S48558 21 x 60 in. 30 in. Leg Brace, UL-rated Electrical
S48572 21 x 60 in. 36 in. Leg Brace, UL-rated Electrical
S41821 36 x 72 in. 34 in. Leg Brace, UL-rated Electrical
S48718 21 x 72 in. 34 in. Leg Brace, UL-rated Electrical
S48643 36 x 60 in. 30 in. Leg Brace, UL-rated Electrical
S48711 21 x 72 in. 30 in. Leg Brace, UL-rated Electrical
S48657 36 x 60 in. 36 in. Leg Brace, UL-rated Electrical
S48650 36 x 60 in. 34 in. Leg Brace, UL-rated Electrical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
Catalog No. S42744 Supplier Diversified Spaces™ Supplier No. P7144K30LE
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces and UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Includes Leg Brace, UL-rated Electrical
Top Style Solid Phenolic
Certifications/Compliance SEFA compliant and MAS Certified Green™
Style Plain Apron
Adjustable No
For Use With (Application) High school and above and with low temperature use
Load Bearing Capacity (English) 500 lb.
Dimensions (L x W) 30 x 60 in. (76.2 x 152.4 cm)
Finish Chemical resistant, earth-friendly UV
Height (English) 30 in.
Height (Metric) 76.2 cm
Load Bearing Capacity (Metric) 226.79 kg
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.