Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Leg Brace Option
SCI_ED

Catalog No. S42232 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 48 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 48 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
42 x 48 in.
42 x 54 in.
42 x 60 in.
Height (English):
30 in.
34 in.
36 in.
Includes:
Leg Brace
40 product options available for selection
Product selection table with 40 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English) Includes
S42232 42 x 54 in. 36 in. Leg Brace
S41909 24 x 72 in. 30 in. Leg Brace
S42108 24 x 60 in. 30 in. Leg Brace
S42122 24 x 60 in. 36 in. Leg Brace
S42227 42 x 54 in. 34 in. Leg Brace
S42285 42 x 60 in. 34 in. Leg Brace
S42290 42 x 60 in. 36 in. Leg Brace
S42743 30 x 60 in. (76.2 x 152.4 cm) 30 in. Leg Brace
S42747 30 x 60 in. (76.2 x 152.4 cm) 34 in. Leg Brace
S47457 24 x 48 in. 30 in. Leg Brace
S47467 24 x 48 in. 36 in. Leg Brace
S47462 24 x 48 in. 34 in. Leg Brace
S47551 42 x 48 in. 34 in. Leg Brace
S47613 30 x 48 in. 34 in. Leg Brace
S47608 30 x 48 in. 30 in. Leg Brace
S47556 42 x 48 in. 36 in. Leg Brace
S47618 30 x 48 in. 36 in. Leg Brace
S47681 30 x 54 in. 30 in. Leg Brace
S47822 30 x 60 in. (76.2 x 152.4 cm) 36 in. Leg Brace
S47695 30 x 54 in. 36 in. Leg Brace
S48063 21 x 28 in. 30 in. Leg Brace
S47968 30 x 72 in. 34 in. Leg Brace
S48077 21 x 28 in. 36 in. Leg Brace
S48070 21 x 28 in. 34 in. Leg Brace
S48155 36 x 48 in. 34 in. Leg Brace
S48148 36 x 48 in. 30 in. Leg Brace
S48227 21 x 54 in. 30 in. Leg Brace
S48234 21 x 54 in. 34 in. Leg Brace
S48312 36 x 54 in. 30 in. Leg Brace
S48319 36 x 54 in. 34 in. Leg Brace
S48430 24 x 54 in. 30 in. Leg Brace
S48444 24 x 54 in. 36 in. Leg Brace
S48326 36 x 54 in. 36 in. Leg Brace
S48437 24 x 54 in. 34 in. Leg Brace
S48564 21 x 60 in. 34 in. Leg Brace
S48571 21 x 60 in. 36 in. Leg Brace
S41813 36 x 72 in. 30 in. Leg Brace
S41820 36 x 72 in. 34 in. Leg Brace
S48656 36 x 60 in. 36 in. Leg Brace
S48717 21 x 72 in. 34 in. Leg Brace
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 40 options available.
40 options
Catalog No. S42232 Supplier Diversified Spaces™ Supplier No. P7804K36L
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces, stretcher, and UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Dimensions (L x W) 42 x 54 in.
Certifications/Compliance SEFA compliant and MAS Certified Green™
Includes Leg Brace
Finish Chemical resistant, earth-friendly UV
Top Style Solid Phenolic
Style Plain Apron
Adjustable No
For Use With (Application) High school and above and with low temperature use
Height (English) 36 in.
Height (Metric) 91.44 cm
Load Bearing Capacity (English) 500 lb.
Load Bearing Capacity (Metric) 226.79 kg
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.