Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Leg Brace, Stretcher and Caster Option
SCI_ED

Catalog No. S42700 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
42 x 48 in.
42 x 54 in.
42 x 60 in.
Height (English):
30 in.
34 in.
36 in.
Includes:
Leg Brace, Stretcher, Caster
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English) Includes
S42700 42 x 48 in. 36 in. Leg Brace, Stretcher, Caster
S41913 24 x 72 in. 30 in. Leg Brace, Stretcher, Caster
S41923 24 x 72 in. 36 in. Leg Brace, Stretcher, Caster
S41920 24 x 72 in. 34 in. Leg Brace, Stretcher, Caster
S42112 24 x 60 in. 30 in. Leg Brace, Stretcher, Caster
S42119 24 x 60 in. 34 in. Leg Brace, Stretcher, Caster
S42230 42 x 54 in. 34 in. Leg Brace, Stretcher, Caster
S42235 42 x 54 in. 36 in. Leg Brace, Stretcher, Caster
S42126 24 x 60 in. 36 in. Leg Brace, Stretcher, Caster
S42288 42 x 60 in. 34 in. Leg Brace, Stretcher, Caster
S42293 42 x 60 in. 36 in. Leg Brace, Stretcher, Caster
S47685 30 x 54 in. 30 in. Leg Brace, Stretcher, Caster
S47549 42 x 48 in. 30 in. Leg Brace, Stretcher, Caster
S47692 30 x 54 in. 34 in. Leg Brace, Stretcher, Caster
S47554 42 x 48 in. 34 in. Leg Brace, Stretcher, Caster
S47699 30 x 54 in. 36 in. Leg Brace, Stretcher, Caster
S47826 30 x 60 in. (76.2 x 152.4 cm) 36 in. Leg Brace, Stretcher, Caster
S48067 21 x 28 in. 30 in. Leg Brace, Stretcher, Caster
S47975 30 x 72 in. 36 in. Leg Brace, Stretcher, Caster
S48152 36 x 48 in. 30 in. Leg Brace, Stretcher, Caster
S48074 21 x 28 in. 34 in. Leg Brace, Stretcher, Caster
S48159 36 x 48 in. 34 in. Leg Brace, Stretcher, Caster
S48081 21 x 28 in. 36 in. Leg Brace, Stretcher, Caster
S48166 36 x 48 in. 36 in. Leg Brace, Stretcher, Caster
S48231 21 x 54 in. 30 in. Leg Brace, Stretcher, Caster
S48245 21 x 54 in. 36 in. Leg Brace, Stretcher, Caster
S48238 21 x 54 in. 34 in. Leg Brace, Stretcher, Caster
S48330 36 x 54 in. 36 in. Leg Brace, Stretcher, Caster
S48568 21 x 60 in. 34 in. Leg Brace, Stretcher, Caster
S48448 24 x 54 in. 36 in. Leg Brace, Stretcher, Caster
S48575 21 x 60 in. 36 in. Leg Brace, Stretcher, Caster
S48561 21 x 60 in. 30 in. Leg Brace, Stretcher, Caster
S48441 24 x 54 in. 34 in. Leg Brace, Stretcher, Caster
S41817 36 x 72 in. 30 in. Leg Brace, Stretcher, Caster
S41824 36 x 72 in. 34 in. Leg Brace, Stretcher, Caster
S42537 36 x 72 in. 36 in. Leg Brace, Stretcher, Caster
S48646 36 x 60 in. 30 in. Leg Brace, Stretcher, Caster
S48660 36 x 60 in. 36 in. Leg Brace, Stretcher, Caster
S48714 21 x 72 in. 30 in. Leg Brace, Stretcher, Caster
S48653 36 x 60 in. 34 in. Leg Brace, Stretcher, Caster
S48721 21 x 72 in. 34 in. Leg Brace, Stretcher, Caster
S48724 21 x 72 in. 36 in. Leg Brace, Stretcher, Caster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
Catalog No. S42700 Supplier Diversified Spaces™ Supplier No. P7114K36SC
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with 3 in. locking swivel casters
  • Includes leg braces, stretcher and casters
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Includes Leg Brace, Stretcher, Caster
Dimensions (L x W) 42 x 48 in.
Height (Metric) 91.44 cm
For Use With (Application) High school and above and with low temperature use
Height (English) 36 in.
Load Bearing Capacity (English) 500 lb.
Finish Chemical resistant, earth-friendly UV
Load Bearing Capacity (Metric) 226.79 kg
Top Style Solid Phenolic
Style Plain Apron
Certifications/Compliance SEFA compliant and MAS Certified Green™
Adjustable No
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.