Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Oak Table with Plain Apron and Solid Phenolic Top: Leg Brace, Stretcher and Electrical Option
SCI_ED

Catalog No. S48442 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (L x W):
21 x 28 in.
21 x 54 in.
21 x 60 in.
21 x 72 in.
24 x 54 in.
24 x 60 in.
24 x 72 in.
30 x 54 in.
30 x 60 in. (76.2 x 152.4 cm)
30 x 72 in.
36 x 48 in.
36 x 54 in.
36 x 60 in.
36 x 72 in.
42 x 48 in.
42 x 54 in.
42 x 60 in.
42 x 72 in.
Height (English):
30 in.
34 in.
36 in.
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Height (English)
S48442 24 x 54 in. 34 in.
S41921 24 x 72 in. 34 in.
S42000 42 x 72 in. 30 in.
S41924 24 x 72 in. 36 in.
S42004 42 x 72 in. 34 in.
S42113 24 x 60 in. 30 in.
S42120 24 x 60 in. 34 in.
S42226 42 x 54 in. 30 in.
S42127 24 x 60 in. 36 in.
S42284 42 x 60 in. 30 in.
S42236 42 x 54 in. 36 in.
S42294 42 x 60 in. 36 in.
S47560 42 x 48 in. 36 in.
S47550 42 x 48 in. 30 in.
S47686 30 x 54 in. 30 in.
S47693 30 x 54 in. 34 in.
S47973 30 x 72 in. 34 in.
S47966 30 x 72 in. 30 in.
S47820 30 x 60 in. (76.2 x 152.4 cm) 34 in.
S47827 30 x 60 in. (76.2 x 152.4 cm) 36 in.
S47976 30 x 72 in. 36 in.
S48068 21 x 28 in. 30 in.
S48082 21 x 28 in. 36 in.
S48153 36 x 48 in. 30 in.
S48075 21 x 28 in. 34 in.
S48167 36 x 48 in. 36 in.
S48160 36 x 48 in. 34 in.
S48232 21 x 54 in. 30 in.
S48239 21 x 54 in. 34 in.
S48317 36 x 54 in. 30 in.
S48324 36 x 54 in. 34 in.
S48435 24 x 54 in. 30 in.
S48331 36 x 54 in. 36 in.
S48449 24 x 54 in. 36 in.
S48562 21 x 60 in. 30 in.
S48569 21 x 60 in. 34 in.
S48576 21 x 60 in. 36 in.
S41828 36 x 72 in. 36 in.
S48715 21 x 72 in. 30 in.
S48647 36 x 60 in. 30 in.
S48661 36 x 60 in. 36 in.
S48722 21 x 72 in. 34 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
Catalog No. S48442 Supplier Diversified Spaces™ Supplier No. P7204K34SE
Add to Cart
Add to Cart

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.

  • 1 in. Phenolic Table Surface
  • 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Includes leg braces, stretcher and UL rated electrical can be hard wired or portable cord connected by certified electricians
  • Limited Lifetime Warranty
  • Made in the USA
  • Minimal assembly required; legs need to be attached
  • Solid Oak Construction

Specifications

Product Type Plain Apron Table
Color Oak
Dimensions (L x W) 24 x 54 in.
Finish Chemical resistant, earth-friendly UV
For Use With (Application) High school and above and with low temperature use
Load Bearing Capacity (English) 500 lb.
Height (Metric) 86.36 cm
Height (English) 34 in.
Top Style Solid Phenolic
Certifications/Compliance SEFA compliant and MAS Certified Green™
Includes Leg Brace, Stretcher and UL rated Electrical
Style Plain Apron
Adjustable No
Load Bearing Capacity (Metric) 226.79 kg
Material Red Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.