Learn More
Description
Oakton Conductivity and TDS Standard is traceable to NIST standard reference materials. Solutions help users calibrate conductivity instruments to ensure accuracy.
- KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60 mL bottle, but included in insert)
- Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60 mL bottle, but included in insert)
- Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
- If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months
- Conductivity options range from 1 μS/cm to 200,000 μS/cm
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | Plastic |
| Conductivity | 12,880 μS |
| Technique | Calibration of Conductivity Instrument |
| Color | Clear |
| pH | 4.7 to 7.8 |
| Boiling Point | 100°C (212°F) |
| For Use With (Equipment) | Conductivity Meter |
| Solubility | Soluble |
| Solution Type | Standard Solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.