Learn More
Oakton™ Water Analysis Accessories
Catalog No. 15500036 Shop All Oakton Products
Change view
Click to view available options
Capacity (Metric):
500 mL
Catalog No. Capacity (Metric)
15-500-036 500 mL
1 options
Catalog No. 15-500-036 Supplier Oakton™ Supplier No. 3564071
Add to Cart
Add to Cart
For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample
Specifications
Capacity (Metric) 500 mL
Description Oakton DO Probe Replacement Electrolyte Solution
For Use With (Equipment) DO 6, DO 100, DO 110, DO 300, any Non-Oakton™ DO Meter
Type Electrolyte Solution
For Use With (Application) DO measurement
Includes Electrolyte Solution
Probe Type DO