Learn More
Oakton™ Double-Junction Waterproof pHTestrs™
Catalog No. 1330101 Shop All Oakton Products
Change view
Click to view available options
Calibration Points:
3
Catalog No. Calibration Points
13-301-01 3
1 options
Catalog No. 13-301-01 Supplier Oakton Supplier No. 3562485
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pocket pH Meters with Double-Junction Sensor
±0.01pH accuracy; Three-point calibration; Model with temperature display available.
  • Replaceable double-junction electrode in thermoplastic body resists effects of solvents and organics
  • Excellent for use in wastewater, chemical, heavy metal, food, Tris buffer, organic and plating bath applications
  • Floatable waterproof and dustproof housing is ideal for outdoor measurements
  • Certifiable to IP67 standards
  • Push-button operation and calibration with HOLD, AUTO-OFF, and error self-diagnostic functions
  • Large (5/16 in.H [8mm]) LCD
  • Automatic buffer recognition
  • Electrode design and increased reference gel volume extend electrode life
  • Glass pH bulb is recessed for protection against breakage
pHTestr 1. For routine measurements. Single-point calibration.
pHTestr 2. Adds automatic temperature calibration from 0° to 50°C (32° to 122°F) and 3-point pH calibration (4.0, 7.0, 10.0).
pHTestr 3. High-accuracy tester, suitable for sulfide applications and most ASTM and EPA test procedures. With same features as pHTestrs 1 and 2, plus 0.01pH resolution, ±0.01pH accuracy, 0.00 to 14.99 pH testing range, and selectable buffer sets (USA or NIST).
pHTestr 3+. Same as pHTestr 3 but with digital temperature readout (range 0° to 50°C [32° to 122°F]) with an accuracy of ±1°C.
Order Info
Dimensions, 6 1/2L x 1 1/2 in. dia. (16.5 x 4cm). CE listed. All models available with or without calibration kit.
Warranty
One year
Specifications
Calibration Points 3