Learn More
Oakton™ PCTSTestr™ Replacement Sensor for WD-35634-60 Oakton PCTSTestr 5
Catalog No. 14311310 Shop All Oakton Products
Change view
Click to view available options
For Use With (Equipment):
Part number 35634-60
Catalog No. For Use With (Equipment)
14-311-310 Part number 35634-60
1 options
Catalog No. 14-311-310 Supplier Oakton™ Supplier No. 3563462
Add to Cart
Add to Cart
Oakton™ PCTSTestr™ Replacement Sensor for WD-35634-60 Oakton PCTSTestr 5 for when electrodes break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample
  • Provides pH and Conductivity/TDS/Salinity measurements
  • Requires the PCTSTestr 5 meter body
Specifications
Description Oakton Replacement Sensor for PCTSTestr 5
Type Replacement Sensor
Warranty 1 Year
Certifications/Compliance CE
For Use With (Equipment) Part number 35634-60