Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Waterproof Big Display pHTestr™ 10 BNC

Catalog No. 13200264 Shop All Oakton Products
Change view
Click to view available options
Range (pH):
-1 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
13-200-264 -1 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-200-264 Supplier Oakton™ Supplier No. 3563414
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Meter, Four 1.5V batteries, instructions

Requires

Without electrode; accepts most pH combination electrodes with BNC connector.

Pocket pH meter with large display; accepts pH combination electrode with BNC connector

  • Microprocessor-based, with accurate readings to 0.1pH
  • Lets you use most any pH combination electrode with BNC connector
  • Push-button three-point calibration
  • Hold function; auto-off; error messages
  • Waterproof, dustproof and floatable; certifiable to IP67 standards

Order Info

Ordering Information Meter dimensions: 6- in.H x 1-1/2 in. dia. (16.5 x 3.8cm). Shipping weight (meter only): 4.5 oz. (125g).

Warranty and Services

One-year warranty.

Specifications

Accuracy (pH) ±0.01
Includes Meter, Four 1.5V batteries, instructions
For Use With (Application) Ideal for dirty water, heavy sulfides, Tris buffers and organics
Measurement Modes pH
Calibration Automatic
Range (pH) -1 to 15
Resolution (pH) 0.01
Calibration Points 1 to 3
Temperature Range (Metric) -5°C to 50°C
Display Type LCD
Backlight Option None
Certifications/Compliance USA, NIST
IP Rating IP67
Warranty 1 Year
Temperature (English) Operating 32.0°F to 122.0°F
Temperature (Metric) Operating 0°C to 50°C
Type pH Tester
Length (Metric) 16.5 cm
Diameter (English) 1.5 in.
Diameter (Metric) 3.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.