Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oakton™ Waterproof PC 450 Portable Multiparameter Meter Kit

Catalog No. 15500143 Shop All Oakton Products
Change view
Click to view available options
Product Type:
Multiparam. Meter Kit, Cert.
Probe Type:
pH and Conductivity/ATC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Probe Type
15-500-143 Multiparam. Meter Kit, Cert. pH and Conductivity/ATC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-500-143 Supplier Oakton™ Supplier No. 3563091

missing translation for 'validMainframeMsg'

Select meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes.

Choose meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes

  • Rugged, IP67 waterproof housing
  • Intuitive, user-friendly icons; electrode status displays pH slope
  • Large, backlit custom LCD to view main measurement simultaneously with temperature
  • Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container (for use with separate pH and conductivity/ATC probes)
  • Calibrate pH with up to six points from USA, NIST, DIN, and custom pH buffer sets
  • Autocal feature for conductivity has one calibration point per range
  • ATC ensures measurement accuracy even in fluctuating temperatures
  • Memory of 500 data sets and USB or RS-232 output for transfer to PC
  • Other features include: ready (stability) indicator, smart averaging, user-selectable °C/°F units, hold function, calibration alarm, battery life-indicator
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Extended battery life or optional universal power adapter
  • Built-in stand with wall mount allows for easy benchtop measurement or storage

Specifications:

  • pH Range: -2.00 to 16.0
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01
  • mV Range: ±2000
  • mV Resolution: 0.1 to 1
  • mV Accuracy: ±0.5
  • Conductivity Range: 0 to 200mS
  • Conductivity Resolution: 0.01μS to 0.1mS
  • Conductivity Accuracy: ±1% full-scale
  • TDS Range: 0 to 200 ppt
  • TDS Resolution: 0.01ppm to 0.1 ppt
  • TDS Accuracy: ±1% full-scale
  • Salinity Range: 0 to 50 ppt
  • Salinity Resolution: 0.1 ppt
  • Salinity Accuracy: ±1%full-scale
  • Temperature Range: -10° to +110°C (17° to 230°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.5°C (±0.9°F)
  • Temperature Compensation: Manual/Automatic
  • Interface: USB, RS232
  • Power: Two AA batteries, optional AC adapter
  • Battery Life: 500 hours
  • Dimensions (L x W x H): 20 x 8 x 5.7 cm (7.9 x 3.12 x 2.25 in.)

Applications:
Ideal for manufacturing floor, field testing, as well as the laboratory.

Includes:

  • Cat. No. 15-500-140: Meter, combination pH/conductivity/temperature probe, calibration solution, two AA batteries and hard carrying case
  • Cat. No. 15-500-141: Meter, combination pH/conductivity/temperature probe, calibration solution, two AA batteries, hard carrying case and NIST-traceable calibration provided by InnoCal
  • Cat. No. 15-500-142: Meter, pH probe, conductivity/temperature cell K=11.0, calibration solution, Grip-clip electrode holder, two AA batteries and hard carrying case
  • Cat. No. 15-500-143: Meter, pH probe, conductivity/temperature cell K=1.0, calibration solution, Grip-clip electrode holder, two AA batteries, hard carrying case and NIST-traceable calibration certificate provided by InnoCal

Order Info

Order optional data cable for USB or RS-232 connectivity to transfer data to a PC separately.

Certifications

CE marked, TUV 3-1, FCC Class A

Warranty and Services

Three years

Specifications

Accuracy (Conductivity) ±1% full scale
Accuracy (mV) ±0.5
Accuracy (pH) ±0.01
Accuracy (Salinity) ±1% full scale
Accuracy (TDS) ±1% full scale
Battery Type 2 AA
Calibration Points (Conductivity) 1 calibration point per range
Calibration Points (pH) Up to to six points from USA, NIST, DIN, and custom pH buffer sets
Certifications/Compliance CE marked, TUV 3-1, FCC Class A
Data Points 150 Sets
Display Type Backlit LCD
IP Rating IP67
Range (Conductivity) 0 to 200 mS
Range (mV) ±2000
Range (pH) -2 to 16
Range (Salinity) 0 to 50 ppt
Range (TDS) 0 to 200 ppt
Resolution (pH) 0.01
Temperature Range (Metric) -10°C to +110°C
For Use With (Application) Ideal for manufacturing floor, field testing, as well as the laboratory.
Includes Meter, pH probe, conductivity/temperature cell K=1.0, calibration solution, Grip-clip electrode holder, two AA batteries, hard carrying case and NIST-traceable calibration certificate provided by InnoCal
Probe Type pH and Conductivity/ATC
Product Type Multiparam. Meter Kit, Cert.
Resolution (Conductivity) 0.01μS to 0.1mS
Resolution (mV) 0.1 to 1
Resolution (Salinity) 0.1 ppt
Resolution (TDS) 0.01ppm to 0.1 ppt
Temperature Accuracy ±0.5°C, ±0.9°F
Temperature Range (English) 17°F to 230°F
Temperature Resolution 0.1
Warranty 3 Years
Length (English) 7.9 in.
Length (Metric) 20 cm
Width (English) 3.12 in.
Width (Metric) 8 cm
Height (English) 2.25 in.
Height (Metric) 5.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.