Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Oakton™ Waterproof pHTestr BNC Electrodes

Catalog No. 13300165 Shop All Oakton Products
Change view
Click to view available options
Junction:
Double
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Junction
13-300-165 Double
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-300-165 Supplier Cole-Parmer™ Supplier No. 3580402X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Outstanding value at an economical cost!

Electrodes have epoxy body, 12 mm dia., in single- or double-junction style (as indicated).

Specifications

Connector Types BNC
Junction Double
Material Epoxy
pH Range 0 to 14.99
Temperature (Metric) Operating 0°C to 50°C
Type pH Electrode
Drift Unknown
For Use With (Application) For dirty water, high-strength solutions, life sciences, Tris buffers
For Use With (Equipment) Any pH meter with a BNC connector
Includes Electrode and BNC connector
pH Relative Accuracy ±0.02
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Range 80°C Max.
Length (Metric) 155 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.