Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535370
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535370 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535370 Supplier Invitrogen™ Supplier No. PA535370
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OAT Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

OAT is the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme in the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAT
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OAT
Gene Accession No. P04181, P04182, P29758
Gene Alias AI194874; GACR; gyrate atrophy; HOGA; OAT; OATASE; OKT; ornithine aminotransferase; ornithine aminotransferase (gyrate atrophy); Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; ornithine-delta-aminotransferase; ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; rOAT; testicular tissue protein Li 130
Gene Symbols OAT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 27-55 amino acids from the N-terminal region of human OAT.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18242, 4942, 64313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.