Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OATP2/OATP8 Monoclonal Antibody (MDQ)
GREENER_CHOICE

Catalog No. MA3935 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3935 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3935 Supplier Invitrogen™ Supplier No. MA3935
  • $596.37
    $461.65 / Each of 1

    Save $134.72

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-935 detects OATP8 from human tissue and cells. This antibody does not detect dog, mouse or rat protein. MA3-935 also detects the OATP2 isoform. MA3-935 has been successfully used in immunohistochemistry, immunofluorescence and immunoprecipitation procedures. Immunofluorescence staining of OATP8 in human liver samples results in a pattern consistent with plasma membrane staining. The MA3-935 immunogen is a synthetic peptide corresponding ot residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2.

The OATPs (organic anion transporter polypeptides) are a family of transport proteins that have been found to mediate the uptake of organic ions into hepatocyte cells. These proteins fall into the solute carrier family 21A (SLC21A), and are expressed in liver, kidney, and brain. These polypeptides are involved in the excretion of potentially toxic compounds and therefore play a role in the overall detoxification system of the body. Unlike other members of the OATP family, OATP2 and 8 have been detected exclusively in the liver, and there is an 80% amino acid sequence homology between these polypeptides. Other names for OATP2 include SLC21A6, OATP-C, and LST-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP2/OATP8
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone MDQ
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene SLCO1B1
Gene Accession No. Q9NPD5, Q9Y6L6
Gene Alias HBLRR; liver-specific organic anion transporter 1; Liver-specific organic anion transporter 2; liver-specific organic anion transporter 3TM13; LST1; LST-1; LST2; LST-2; LST3; LST-3TM13; OATP1B1; OATP1B3; OATP2; OATP-2; OATP8; OATP-8; OATPC; OATP-C; Organic anion transporter 8; organic anion transporter LST-3c; organic anion-transporting polypeptide 8; SLC21A6; SLC21A8; SLCO1B1; SLCO1B3; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; solute carrier family 21 (organic anion transporter), member 8; Solute carrier family 21 member 6; Solute carrier family 21 member 8; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family member 1B3; solute carrier organic anion transporter family, member 1B1; solute carrier organic anion transporter family, member 1B3
Gene Symbols SLCO1B1, SLCO1B3
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10599, 28234
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.