Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OAZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552662 Supplier Invitrogen™ Supplier No. PA552662
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSRLTDAKRI NWRTVLSGGS LYIEIPGGAL PEGSKDSFAV LLEFAEEQLR ADHVFICFHK NREDRAALLR TFSFLGFEIV RPGHPLVPKR PDACFMAYTF ERESSGEEEE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

OAZ1 catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. This protein play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein expression is auto-regulated by polyamine-enhanced translational frameshifting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAZ1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OAZ1
Gene Accession No. P54368
Gene Alias Antizyme; antizyme 1; AZ1; AZ-1; AZI; OAZ; OAZ1; ODCAC; ODC-Az; Ornithine decarboxylase antizyme 1
Gene Symbols OAZ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OAZ1. Recombinant protein control fragment (Product #RP-89601).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4946
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.