Learn More
Santa Cruz Biotechnology OBFC1 Antibody (E-10): sc-376450, 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC376450
Encompass
OBFC1 Antibody (E-10) is a mouse monoclonal IgG1 κ OBFC1 antibody, cited in 5 publications, provided at 200 µg/mlraised against amino acids 8-307 mapping at the N-terminus of OBFC1 of mouse originOBFC1 Antibody (E-10) is recommended for detection of OBFC1 of mouse, rat and human origin by WB, IP, IF and ELISA
Catalog No. NC2752807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.