Learn More
Cayman Chemical ObIdoxImechlorIde 250mg
SDP
Supplier:  Cayman Chemical 36587250 MG
Encompass_Preferred
An NSAID inhibits ragweed- A23187- or concanavalin A-induced histamine release in isolated human leukocytes at 0 5 mM inhibits carrageenan-induced paw edema in rats (ED50 67 mg/kg)
Catalog No. 50-290-9486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.