Learn More
Invitrogen™ OBP Polyclonal Antibody

Description
OBP Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | OBP |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | OBP |
| Gene Accession No. | P07435 |
| Gene Alias | 1110006M06Rik; AI462538; Obph1; ORP5; ORP-5; Osbp2; Osbpl5; OSBP-related protein 5; oxysterol binding protein 2; oxysterol binding protein homolog 1; oxysterol binding protein-like 5; oxysterol-binding protein homolog 1; oxysterol-binding protein-like protein 5; oxysterol-binding protein-related protein 5 |
| Gene Symbols | OBP, Osbpl5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.