Learn More
OccuNomix™ Cap Liners
Multiple styles from which to choose
Supplier:  OccuNomix™ SS550X
 View more versions of this product
Catalog No. 19-520-381
This item is not returnable. View return policy
Description
Description
  • Knitted Tube Liner: 100% polyester knit fits snugly over hardhat; folds up on outside of hat when not needed. Blocks wind tunneling inside crown of hat.
  • Heavy-Duty Shoulder-Length Liner: Shoulder-length cotton twill shell with sherpa lining. Two layers, warming pack pockets, elastic for snug fit, and insulated ear barrier.
  • Value Sherpa Liner: Shoulder length cotton twill shell with Sherpa lining.
  • Mid Length Liner: Single washed FR treated cotton outer shell with poly fleece lining. Two layers, warming pack pockets, reflective strip, elastic for snug fit and insulated ear barrier.
  • Shoulder Length Two-Way Liner: 100% cotton top with polyester-fleece lining; removable 100% polyester bottom. Fleece and polyester are flame resistant.
  • Extra-Long Liner with Removable Mouthpiece: Two layers with navy shell and red lining with extra-long neck length.
  • Three-Layer Shoulder-Length Liner: 100% cotton twill shell with Thinsulate™ thermal middle layer and heavyweight polyester-fleece lining.
  • Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.