Learn More
OccuNomix™ Embossed Rubber Back Anti-Vibration Gloves with VEP Padding
Shop All Occunomix International Inc Products
Description
- Premium leather palm
- 4-way stretch spandex back
- Visco elastic polymer padding
- Hook & Loop wrist closure
- Terry cloth thumb to wipe away sweat
Specifications
Specifications
| Back Material | Spandex |
| Color | Black |
| Hand | Specific |
| Material | Synthetic Leather |
| Product Type | Anti-vibration Gloves |
| Size | X-Large |
| For Use With (Application) | Medium to Heavy Industrial Power Tools |
| Glove Cuff Style | Hook and Loop |
| Series | 426 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.