Learn More
OccuNomix™ Half-Finger Terry Cloth Anti-Vibration Gloves
Shop All Occunomix International Inc Products
Description
- VEP gel padding covers the palm, index finger, and lower thumb
- Optimally suited for impact power tools, sanders, and small power tools
- Hook and loop closure
Specifications
Specifications
| Color | Blue, Gray |
| Hand | Specific |
| Material | Terrycloth |
| Product Type | Anti-vibration Gloves |
| Size | X-Large |
| For Use With (Application) | Power Tools and Sanders |
| Glove Cuff Style | Hook and Loop |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.