Learn More
OccuNomix™ MiraCool™ Bandanas
Lightweight neck bandanas with cooling power and the comfort of cotton
Supplier:  OccuNomix™ 940018
 View more versions of this product
Catalog No. 19-816-246
Only null left
Add to Cart
Description
Description
  • 100 percent cotton with cooling crystals
  • Works without freezing or ice
  • Simply soak in cold water for 10 to 20 minutes
  • Stay hydrated for days
  • Reusable
  • Triple washed to prevent fabric bleeding
  • Flame-resistant, arc-rated, treated cotton (treatment lasts for up to 50 washes)
Specifications
Specifications
Cotton
Universal
Tie
Cooling Bandana
Navy
Cooling Crystals
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.