Learn More
OccuNomix™ Terry Toppers™ Snap-On Sweatbands for Hard Hats
Shop All Occunomix International Inc Products
Description
Installs easily with snap-on application and helps hard hat fit more securely
- Constructed from 100% super-absorbent soft terry cotton
- Offers plush, not scratchy, texture
- Absorbs without needing to be washed first
- No foam middle
- One size fits all
- Machine washable
- Available in navy or beige
Specifications
Specifications
| Color | Beige |
| For Use With (Equipment) | Hard Hats |
| Finish | Textured |
| Material | Terrycloth |
| Product Type | Snap-on Sweatband |
| Size | Universal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.