Learn More
Apexbio Technology LLC KT 5823 500UG
SDP
Supplier:  Apexbio Technology LLC B66730.5
Encompass_Preferred
KT 5823 (CAS 126643-37-6) is a selective inhibitor of protein kinase G (PKG) By specifically inhibiting PKG-mediated phosphorylation pathways KT 5823 serves as an important pharmacological tool in biomedical research for dissecting the role of PKG signaling in various physiological and pathological contexts Researchers utilize KT 5823 extensively to investigate PKG-dependent processes including vascular tone regulation platelet aggregation neuronal signaling and smooth muscle function
Catalog No. 50-000-61218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.