Learn More
Apexbio Technology LLC Suramin hexasodium salt(Synonyms: Suramin sodium, Germanin, Naganol, Moranyl, Bayer 205, Antrypol, Fourneau 309), 10mg, CAS: 129-46-4.
SDP
Supplier:  Apexbio Technology LLC B675210
Encompass_Preferred
Suramin hexasodium salt (CAS 129-46-4) is a polysulfonated naphthylurea derivative that competitively inhibits DNA topoisomerase II an enzyme essential for managing DNA supercoiling and topology during replication and transcription with an IC50 of approximately 5 M Suramin selectively inhibits cellular DNA synthesis and proliferation notably blocking SV40 DNA replication entirely at a concentration of 40 M in HeLa cells The compound demonstrates differential inhibitory effects towards DNA polymerases and (IC50 of 8 M and 36 M respectively) Additionally suramin significantly inhibits bacterial RecA-mediated ATPase activity strand exchange processes and proteolytic cleavage of the LexA transcriptional repressor In preclinical models of pulmonary hypertension suramin reduces vascular remodeling inflammation and pulmonary artery pressure reflecting its utility in cellular proliferation enzyme function and disease model research
Catalog No. 50-000-61280
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.