Learn More
Apexbio Technology LLC A 205804 10MG
SDP
Supplier:  Apexbio Technology LLC A866210
Encompass_Preferred
A 205804 (CAS 251992-66-2) is a selective inhibitor targeting the expression of adhesion molecules E-selectin and ICAM-1 exhibiting IC50 values of 20 nM and 25 nM respectively E-selectin and ICAM-1 mediate endothelial-leukocyte interactions essential in inflammatory responses In human endothelial cell assays (e g HUVECs) A 205804 reduces ICAM-1 and E-selectin expression thus attenuating leukocyte-endothelial adhesion and migration Mechanistically A 205804 translocates to the nucleus binding non-covalently to large ( 650 kDa) molecular targets A 205804 serves as a research tool for studying inflammatory processes and adhesion-associated diseases
Catalog No. 50-000-60073
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.