Learn More
*HELICA BIOSYSTEMS OCHRATOXIN A/COFFEE/COCOA
Supplier:  *HELICA BIOSYSTEMS 961OCH01COF
Encompass_Preferred
Immunochemical Assay Kits; Food Safety; Mycotoxin Detection Assay; ELISA Test Kit, Qualitative Ochratoxin A detection, 96-strip microwell plate based assay, Results in 1.5 hours, One-year stability; Excellent reproducibility
Catalog No. 50-980-594
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.