Learn More
Sigma Aldrich Fine Chemicals Biosciences MS PhosphoMix 1 Light Phos
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MSP1L5X1VL
Encompass
The MS PhosphoMix line of products allows for the testing of the strengths and weaknesses of phosphopeptide sample processing mass spectrometry analysis and instrument configurations. The mixes are produced from synthetic phosphopeptides with sequences derived from naturally occurring peptides as identified by Mann et al. in HeLa cells. Because the sequences are derived from mammalian cells many natural phosphorylation motifs such as those that present an abundance of proline are represented. Additionally the phosphopeptide distribution in each mix has been chosen to present a broad range of characteristics including ionizability LC retention time charge state and isoelectric point. finally PhosphoMix-1 2 and 3 were designed in a complementary fashion as highlighted on the following page. for example all three mixes contain peptides of the same sequence with different sites of phosphorylation.Each of the three phosphopeptides mixes are available in their naturally occurri
Catalog No. 50-291-4203
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.