Learn More
*HELICA BIOSYSTEMS OCHRATOXIN (QUANTITATIVE)/WINE
Supplier:  *HELICA BIOSYSTEMS 981OCH01ALC
Encompass_Preferred
Immunochemical Assay Kits; Food Safety; Mycotoxin Detection Assay; ELISA Test Kit, Quantitative Ochratoxin (wine) Detection, 96-strip microwell plate based assay, Results in 1 hour, One-year stability; Excellent reproducibility
Catalog No. 50-980-592
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.