Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCIAD1 Mouse anti-Human, Mouse, Rat, Clone: 1C10C3, Proteintech
SDP

Catalog No. 501737114 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7114 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7114 Supplier Proteintech Group Inc Supplier No. 666981IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.

Specifications

Antigen OCIAD1
Applications Western Blot
Classification Monoclonal
Clone 1C10C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OCIAD1
Gene Accession No. Q5XIG4, Q9CRD0, Q9NX40
Gene Alias Asrij, FLJ20455, OCIA, OCIA domain containing 1, OCIAD1, TPA018
Gene Symbols Ociad1
Host Species Mouse
Immunogen OCIAD1 Fusion Protein Ag9977
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289590, 54940, 68095
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.