Learn More
Medchemexpress LLC N-methyl-1-[4-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclohexyl]methanesulfonamide | 1208319-26-9 | MFCD25976611 | 99.6% | C15H23N5O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1357710MG
Encompass_Preferred
Oclacitinib is a research-grade small-molecule Janus kinase (JAK) inhibitor most potent against JAK1 (IC50 10 nM). It is provided as a high-purity powder for preclinical and biochemical studies of cytokine-mediated signaling, allergic responses, and inflammatory pathways. Documentation such as a datasheet and certificate of analysis are available to support experimental use.
  • Selective JAK1 inhibition with IC50 10 nM.
  • Suitable for cellular and biochemical assays probing cytokine signaling.
  • High purity (≈99.6%) for reproducible results.
  • Supplied as a powder with recommended long-term storage at -20°C.
  • Datasheet and certificate of analysis available for quality assurance.
Catalog No. 50-002-60824