Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501559525
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9525 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9525 Supplier Bethyl Laboratories, Inc Supplier No. A301717AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Pig

Oct1 or POU2F1 (Pou domain, class 2, transcription factor 1) is a transcription factor that binds to the octamer motif (5′-ATTTGCAT-3′) and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR, and PGR and interacts with NR3C1, AR, and PGR. Oct1 is ubiquitous and belongs to the POU transcription factor family class 2 subfamily. Oct1 contains 1 homeobox domain.

Specifications

Antigen OCT1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene POU2F1
Gene Accession No. P14859
Gene Alias OCT1, OTF1
Gene Symbols POU2F1
Host Species Rabbit
Immunogen Between 693 and 743
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5451
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.