Learn More
MBL International Corporation Anti-Oct3/4 (Mouse) pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation PM048
Encompass_Preferred
Clonality:Polyclonal; Application:ICC|IHC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Human|Mouse; Size:100 µl; Target:Oct3/4; Polyclonal Antibody of 100 targeting Oct3/4 for ICC, IHC, IP, WB.Oct3/4 Polyclonal Antibody.
Catalog No. 50-507-60
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.