Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCT4 Monoclonal Antibody (9B7), DyLight™ 488
GREENER_CHOICE

Catalog No. MA1104D488 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1104D488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1104D488 Supplier Invitrogen™ Supplier No. MA1104D488
  • $555.50
    $417.65 / Each of 1

    Save $137.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-104-D488 detects Oct4/Pou5F1 in human iPS samples. MA1-104-D488 has been successfully used in ICC/IF applications.

POU5F1, also commonly known as Oct-4, is a maternally expressed octamer-binding protein that was the first transcription factor described for the early stages of development. The role of POU5F1 in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing POU5F1, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT4
Applications Immunocytochemistry
Classification Monoclonal
Clone 9B7
Concentration 1 mg/mL
Conjugate DyLight 488
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene POU5F1
Gene Accession No. Q01860
Gene Alias 2-Oct; 3-Oct; 4-Oct; cb197; chunp6868; DADB-104B20.2; etID49452.21; gp9; gp-9; HGNC:9221; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; MGC22487; NF-A3; Oct2; Oct-2; Oct2a; Oct2b; OCT3; Oct-3; Oct3/4; Oct-3/4; OCT4; Oct-4; Octamer-binding protein 2; octamer-binding protein 3; Octamer-binding protein 4; octamer-binding transcription factor 2; Octamer-binding transcription factor 3; OTF2; OTF-2; OTF3; Otf-3; Otf3g; Otf3-rs7; Otf4; Otf-4; OTTHUMP00000221150; OTTHUMP00000221151; POU class 2 homeobox 2; POU class 5 homeobox 1; POU class 5 homeobox 1 transcript variant OCT4B1; POU class 5 homeobox 1 transcript variant OCT4B2; POU class 5 homeobox 1 transcript variant OCT4B3; POU class 5 homeobox 1 transcript variant OCT4B4; POU class 5 homeobox 1 transcript variant OCT4B5; POU class 5 homeobox 1 transcript variant OCT4B6; POU domain class 5 transcription factor 1; POU domain gene 2; POU domain protein 2; POU domain transcription factor OCT4; POU domain, class 2, transcription factor 2; POU domain, class 5, transcription factor 1; POU domain, class 5, transcription factor 3; pou2; pou-2; POU2F2; Pou5f1; pou5f1 protein; pou5f3; POU-type homeodomain-containing DNA-binding protein; spg; spg/pou2; spiel-ohne-grenzen; spiel-ohne-grenzen/pou2; wu:fd18d06
Gene Symbols POU5F1
Host Species Mouse
Immunogen Full length human recombinant protein of human POU5F1 produced in E.coli.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.