Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT4 Mouse anti-Human, HRP, Clone: 1C4B6, Proteintech
SDP

Catalog No. 501737236 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7236 Supplier Proteintech Group Inc Supplier No. HRP60242100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Oct4 is a maternally expressed octamer-binding protein that was the first transcription factor described for the early stages of development. Oct4 is one of the four Yamaka factors, necessary for embryonic stem cell pluripotency. Pluripotent stem (iPS) cells, can be created by expressing Oct4 and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Oct4, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. A translocation of this gene with the Ewing′s sarcoma gene, t(6;22)(p21;q12), has been linked to tumor formation.

Specifications

Antigen OCT4
Applications Western Blot
Classification Monoclonal
Clone 1C4B6
Concentration 0.67 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POU5F1
Gene Accession No. Q01860
Gene Alias Oct 3, Oct 4, OCT3, OCT3/4, OCT4, Octamer binding protein 3, Octamer binding protein 4, OTF 3, OTF3, OTF4, POU class 5 homeobox 1, POU5F1
Gene Symbols POU5F1
Host Species Mouse
Immunogen OCT4 Fusion Protein Ag1794
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.