Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecanamide, MP Biomedicals

Catalog No. ICN204422 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204422 Supplier MP Biomedicals, Inc Supplier No. 0520442225

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124-26-5
Molecular Formula C18H37NO
Molecular Weight (g/mol) 283.50
MDL Number MFCD00008038
InChI Key LYRFLYHAGKPMFH-UHFFFAOYSA-N
Synonym stearamide, stearylamide, octadecylamide, stearoylamide, stearoylamine, octadecamide, stearic acid amide, stearic amide, kemamide s, adogen 42
PubChem CID 31292
ChEBI CHEBI:34900
IUPAC Name octadecanamide
SMILES CCCCCCCCCCCCCCCCCC(N)=O

Specifications

Quantity 100 g
Formula Weight 283.5
Chemical Name or Material Octadecanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.