Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A0675250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0675250G 250 g
A067525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0675250G Supplier TCI America Supplier No. A0675250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 822-23-1
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00043649
InChI Key OIZXRZCQJDXPFO-UHFFFAOYSA-N
Synonym Acetic Acid Octadecyl Ester, Acetic Acid Stearyl Ester, Stearyl Acetate
PubChem CID 69968
IUPAC Name octadecyl acetate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C

Specifications

Color White
Quantity 250 g
Formula Weight 312.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octadecyl Acetate
EINECSNumber (2)-0735
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.