Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T14715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14715G Supplier TCI America Supplier No. T14715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3386-32-1
Molecular Formula C25H44O3S
Molecular Weight (g/mol) 424.684
MDL Number MFCD00191588
InChI Key IZSBVVGANRHKEE-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid Octadecyl Ester, Stearyl p-Toluenesulfonate, p-Toluenesulfonic Acid Stearyl Ester
PubChem CID 229551
IUPAC Name octadecyl 4-methylbenzenesulfonate
SMILES CCCCCCCCCCCCCCCCCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 424.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octadecyl p-Toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.