Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecyl Vinyl Ether 85.0+%, TCI America™
SDP

Catalog No. O001925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O001925ML 25 mL
O0019500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O001925ML Supplier TCI America Supplier No. O001925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 930-02-9
Molecular Formula C20H40O
Molecular Weight (g/mol) 296.539
MDL Number MFCD00026686
InChI Key QJJDJWUCRAPCOL-UHFFFAOYSA-N
Synonym Stearyl Vinyl Ether
PubChem CID 13585
IUPAC Name 1-ethenoxyoctadecane
SMILES CCCCCCCCCCCCCCCCCCOC=C

Specifications

Color White-Yellow
Quantity 25 mL
Formula Weight 296.54
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Octadecyl Vinyl Ether
EINECSNumber (2)-0372&(2)-0575
RTECSNumber RG0300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.