Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. O016125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016125G 25 g
O01615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O016125G Supplier TCI America Supplier No. O016125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4445-06-1
Molecular Formula C24H48
Molecular Weight (g/mol) 336.65
MDL Number MFCD00058953
InChI Key MWWVNZNVTGBKQO-UHFFFAOYSA-N
Synonym 1-Cyclohexyloctadecane, Stearylcyclohexane
PubChem CID 20514
IUPAC Name octadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCCCC1CCCCC1

Specifications

Melting Point 43°C
Color White
Boiling Point 409°C
Quantity 25 g
Formula Weight 336.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Octadecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.