Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octaethylene Glycol Monomethyl Ether 96.0+%, TCI America™
SDP

Catalog No. O02961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02961G 1 g
O02965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02961G Supplier TCI America Supplier No. O02961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25990-96-9
Molecular Formula C17H36O9
Molecular Weight (g/mol) 384.47
MDL Number MFCD06797152
InChI Key SZGNWRSFHADOMY-UHFFFAOYSA-N
Synonym mPEG8-Alcohol
PubChem CID 526558
IUPAC Name 2,5,8,11,14,17,20,23-octaoxapentacosan-25-ol
SMILES COCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Boiling Point 216°C
Quantity 1 g
Formula Weight 384.47
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Octaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.