Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluoro-1,4-diiodobutane (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. D232925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D232925G 25 g
D23295G 5 g
D232910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D232925G Supplier TCI America Supplier No. D232925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375-50-8
Molecular Formula C4F8I2
Molecular Weight (g/mol) 453.84
MDL Number MFCD00042263
InChI Key JILAKKYYZPDQBE-UHFFFAOYSA-N
Synonym 1,4-diiodooctafluorobutane, octafluoro-1,4-diiodobutane, 1,4-diiodoperfluorobutane, perfluoro-1,4-diiodobutane, 1,4-diiodo octafluorobutane, butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo, acmc-1aigi, i cf2 4i, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
PubChem CID 78993
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Boiling Point 63°C
Quantity 25 g
Formula Weight 453.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octafluoro-1,4-diiodobutane (stabilized with Copper chip)
EINECSNumber (2)-4014
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.