Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluorotoluene 98.0+%, TCI America™
SDP

Catalog No. P085625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P085625G 25 g
P08565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P085625G Supplier TCI America Supplier No. P085625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 434-64-0
Molecular Formula C7F8
Molecular Weight (g/mol) 236.06
MDL Number MFCD00000375
InChI Key USPWUOFNOTUBAD-UHFFFAOYSA-N
Synonym octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
PubChem CID 9906
IUPAC Name 1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
UN Number 1993
Formula Weight 236.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octafluorotoluene
RTECSNumber XT3530000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.