Learn More
Dynalon Labware™ Octagonal Stirring Bar Kit
SCI_ED
PTFE octagonal stirring bars feature ALNICO V magnetic cores which are one of the most powerful permanent magnet alloys available
Catalog No. S29161
Only null left
Add to Cart
Description
Description
Contains octagonal stirring bars shown below. Designed to meet your requirements for a variety of sizes. The 16 piece stir bar kit is packed in a reusable box.16 Stir Bar Kit packed in reusable box contains:(2) each 5/16 x 1/2” (8x13mm)
Specifications
Specifications
0.28 in., 0.4 in., 0.52 in., 0.63 in., 0.95 in., 1.5 in., 2.01 in., 2.52 in.
7, 10, 13, 16, 25, 38, 51, 64 mm
Octagon
0.22kg
White
2 x 7mm, 3 x 10mm, 8 x 13mm, 8 x 16mm, 9.5 x 25mm, 9.5 x 38mm, 9.5 x 51mm, 9.5 x 64mm
Autoclavable
2, 3, 8, 9.5 mm
16
PTFE
0.08 in., 0.12 in., 0.315 in., 0.38 in.
Stirring
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.