Learn More
Sartorius Octet 384TW Plates
Supplier:  Sartorius 185166
Encompass
The Octet 384TW microplate is a black polypropylene 384-well tilted-bottom plate designed for Sartorius’ Octet family of label-free biomolecular interaction analysis instruments. It can be used on the Octet RH96 and RH16 instruments all of which accommodate the 384-well microplate format. The 384TW microplate enables the use of samples as small as 40 μL in Octet assays and reduces variability in the background signal thus improving assay sensitivity particularly beneficial for peptide small molecule and fragment analysis.Note: This product is not returnable. If you have any issues with this product please contact your Sartorius sales representative for assistance.
Catalog No. 50-289-1584