Learn More
Sartorius Sartorius Octet AAVX Biosensors, Rapid AAV Capsid Quantitation, Label-Free Measurement, 5 Trays
Catalog No. NC2389522 Shop All Sartorius Products
Encompass
The Sartorius Octet AAVX Biosensors provide a rapid and label-free solution for quantifying Adeno-Associated Virus (AAV) capsids using Bio-Layer Interferometry (BLI). Designed for efficiency, these biosensors allow measurement in both crude and purified samples, featuring a regeneration capability up to 20 cycles. This streamlined approach enhances process development and quality control in biopharmaceutical applications, making it superior over traditional ELISA methods.
Catalog No. NC2389522 Supplier Sartorius Supplier No. 185161
Add to Cart
Add to Cart
Request a Quote
The Sartorius Octet AAVX Biosensors provide a rapid and label-free solution for quantifying Adeno-Associated Virus (AAV) capsids using Bio-Layer Interferometry (BLI). Designed for efficiency, these biosensors allow measurement in both crude and purified samples, featuring a regeneration capability up to 20 cycles. This streamlined approach enhances process development and quality control in biopharmaceutical applications, making it superior over traditional ELISA methods.