Learn More
Sartorius Sartorius Octet AHC2 Biosensors, Advanced IgG Fc Capture, High Sensitivity, 5 pk
Catalog No. NC3369988 Shop All Sartorius Products
Encompass
The Sartorius Octet AHC2 Biosensors are advanced second-generation biosensors designed for efficient quantitation and kinetic characterization of human IgG antibodies. They enhance analytic sensitivity and dynamic range, offering 10-20 reuses with a low pH regeneration protocol, making them cost-effective for high-throughput applications. This performance, combined with high specificity over all human IgG subclasses and compatibility with both crude and purified samples, differentiates them in the market.
Catalog No. NC3369988 Supplier Sartorius Supplier No. 185143
Add to Cart
Add to Cart
Request a Quote
The Sartorius Octet AHC2 Biosensors are advanced second-generation biosensors designed for efficient quantitation and kinetic characterization of human IgG antibodies. They enhance analytic sensitivity and dynamic range, offering 10-20 reuses with a low pH regeneration protocol, making them cost-effective for high-throughput applications. This performance, combined with high specificity over all human IgG subclasses and compatibility with both crude and purified samples, differentiates them in the market.