Learn More
Sartorius Octet AHC2 Biosensors / Case
Supplier:  Sartorius 185144
Encompass
Octet AHC2 Biosensors. Trays of 96 biosensors coated with anti-human IgG for both quantitation and kinetic analysis of human IgGs and IgG-derived Fc-fusion proteins.
Catalog No. 50-289-1658
Add to Cart
Request a Quote