Learn More
Sartorius Sartorius Octet AMC2 Biosensors, High Sensitivity Kinetic Assays, Reusable Up To 10 Times, 5 Pack
Catalog No. NC3369985 Shop All Sartorius Products
Encompass
The Sartorius Octet AMC2 Biosensors are advanced tools for the high-sensitivity quantification and kinetic characterization of murine and rat IgG antibodies. They provide up to a 3-fold increase in ligand binding capacity and can accurately measure IgG proteins in a wide range, facilitating efficient and cost-effective research applications. With their ability to regenerate up to 10 times, these biosensors are ideal for high-throughput environments like hybridoma screening and quality control.
Catalog No. NC3369985 Supplier Sartorius Supplier No. 185164
Add to Cart
Add to Cart
Request a Quote
The Sartorius Octet AMC2 Biosensors are advanced tools for the high-sensitivity quantification and kinetic characterization of murine and rat IgG antibodies. They provide up to a 3-fold increase in ligand binding capacity and can accurately measure IgG proteins in a wide range, facilitating efficient and cost-effective research applications. With their ability to regenerate up to 10 times, these biosensors are ideal for high-throughput environments like hybridoma screening and quality control.